Läs mer
1,4-Butanediol dimethacrylate 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifikationer
Specifikationer
| Kemiskt namn eller material | 1,4-Butanediol Dimethacrylate |
| CAS | 2082-81-7 |
| Analyt- eller komponentnamn | 1,4-Butanediol Dimethacrylate |
| Frakt skick | Room Temperature |
| Molekylformel | C12 H18 O4 |
| Koncentration | 100 μg/mL |
| Certifikat och efterlevnad | ISO 17034 |
| Synonym | 2-Propenoic acid, 2-methyl-, 1,4-butanediyl ester (9CI), Methacrylic acid, tetramethylene ester (6CI,7CI,8CI), 1,4-Butanediol, dimethacrylate (8CI), 1,4-Butylene dimethacrylate, 1,4-Butylene glycol dimethacrylate, 1,4-Tetramethylene dimethacrylate, BDDMA, BDMA, Butanediol dimethacrylate, DMM 4, Light Ester 1.4BG, Mhoromer MFM 405, NK Ester BD, Oligotetramethylene glycol dimethacrylate, SR 214, SR 214B, SR 214E, Sartomer 214, Tetramethylene dimethacrylate, Tetramethylene glycol dimethacrylate, Tetramethylene methacrylate, Visiomer 1,4-BDDMA, n-Butane-1,4-diyl dimethacrylate, 2-Propenoic acid, 2-methyl-, 1,1'-(1,4-butanediyl) ester |
| InChI formel | InChI=1S/C12H18O4/c1-9(2)11(13)15-7-5-6-8-16-12(14)10(3)4/h1,3,5-8H2,2,4H3 |
| LEDER | CC(=C)C(=O)OCCCCOC(=O)C(=C)C |
| Visa mer |