Läs mer
1,4-Butanediol Dimethylacrylate, TRC
Produktkod. 30179931
Change view
Kvantitet:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179931 1 g
30179827 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179931 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB7100331G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2082-81-7
Molekylformel C12 H18 O4
Molekylvikt (g/mol) 226.2689
Synonym 2-Propenoic acid, 2-methyl-, 1,4-butanediyl ester (9CI), Methacrylic acid, tetramethylene ester (6CI,7CI,8CI), 1,4-Butanediol, dimethacrylate (8CI), 1,4-Butylene dimethacrylate, 1,4-Butylene glycol dimethacrylate, 1,4-Tetramethylene dimethacrylate, BDDMA, BDMA, Butanediol dimethacrylate, DMM 4, Light Ester 1.4BG, Mhoromer MFM 405, NK Ester BD, Oligotetramethylene glycol dimethacrylate, SR 214, SR 214B, SR 214E, Sartomer 214, Tetramethylene dimethacrylate, Tetramethylene glycol dimethacrylate, Tetramethylene methacrylate, Visiomer 1,4-BDDMA, n-Butane-1,4-diyl dimethacrylate, 1,4-Butanediol Dimethacrylate, 2-Propenoic acid, 2-methyl-, 1,1'-(1,4-butanediyl) ester
IUPAC-namn 4-(2-methylprop-2-enoyloxy)butyl 2-methylprop-2-enoate
LEDER CC(=C)C(=O)OCCCCOC(=O)C(=C)C
Specifikationer
Kemiskt namn eller material 1,4-Butanediol Dimethacrylate
Kvantitet 1 g
InChI formel InChI=1S/C12H18O4/c1-9(2)11(13)15-7-5-6-8-16-12(14)10(3)4/h1,3,5-8H2,2,4H3
Formel vikt 226.1205