Läs mer
1-(4-Chloro-2-(2-fluorobenzoyl)phenyl)-2-methyl-1H-imidazole-5-carboxylic Acid, TRC
Produktkod. 30356057
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30356057 10 mg
30364248 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30356057 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC46750010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-[4-Chloro-2-(2-fluorobenzoyl)phenyl]-2-methyl-1H-imidazole-5-carboxylic Acid
CAS 151921-06-1
Molekylformel C18 H12 Cl F N2 O3
Kvantitet 10 mg
Synonym 1-[4-Chloro-2-(2-fluorobenzoyl)phenyl]-2-methyl-1H-imidazole-5-carboxylic Acid, 3-[4-Chloro-2-(2-fluorobenzoyl)phenyl]-2-methyl-3H-imidazole-4-carboxylic Acid
InChI formel InChI=1S/C18H12ClFN2O3/c1-10-21-9-16(18(24)25)22(10)15-7-6-11(19)8-13(15)17(23)12-4-2-3-5-14(12)20/h2-9H,1H3,(H,24,25)
LEDER Cc1ncc(C(=O)O)n1c2ccc(Cl)cc2C(=O)c3ccccc3F
IUPAC-namn 3-[4-chloro-2-(2-fluorobenzoyl)phenyl]-2-methylimidazole-4-carboxylic acid
Molekylvikt (g/mol) 358.75
Formel vikt 358.052