Läs mer
1-(4-Chloro-2-hydroxyphenyl)-1,2-dihydro-3H-pyrazol-3-one, TRC
Produktkod. 30356356
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30356356 250 mg
30364675 50 mg
30364110 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30356356 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC423995250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 512165-97-8
Molekylformel C9 H7 Cl N2 O2
Molekylvikt (g/mol) 210.617
Synonym 3H-Pyrazol-3-one, 1-(4-chloro-2-hydroxyphenyl)-1,2-dihydro-, BF 500-8, 500-M85, M500F085
IUPAC-namn 1-(4-chloro-2-hydroxyphenyl)pyrazol-3-ol
LEDER Oc1ccn(n1)c2ccc(Cl)cc2O
Specifikationer
Kemiskt namn eller material 1-(4-Chloro-2-hydroxyphenyl)-3-hydroxypyrazole
Kvantitet 250 mg
InChI formel InChI=1S/C9H7ClN2O2/c10-6-1-2-7(8(13)5-6)12-4-3-9(14)11-12/h1-5,13H,(H,11,14)
Formel vikt 210.0196