missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chloro-2-nitrophenyl)azepane, 96%, Thermo Scientific™

Catalog Number p-7055367
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
40832-89-1
1-(4-Chloro-2-nitrophenyl)hexamethyleneimine
This item is not returnable. View return policy
40832-89-1
1-(4-Chloro-2-nitrophenyl)hexamethyleneimine

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3259
Beilstein 219458
Formula Weight 254.71
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 1-(4-Chloro-2-nitrophenyl)azepane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'