missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chloro-2-nitrophenyl)pyrrolidine, 97%, Thermo Scientific™

Catalog Number p-7055071
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
41173-36-8
C10H11ClN2O2
226.66
QATZMIJXHKOELL-UHFFFAOYSA-N
1-4-chloro-2-nitrophenyl pyrrolidine, 1-4-chloro-2-nitro-phenyl pyrrolidine, n-4-chloro-2-nitrophenyl pyrrolidine, 4-chloro-2-nitrophenyl pyrrolidine, qatzmijxhkoell-uhfffaoysa, 1-4-chloro-2-nitro-phenyl-pyrrolidine, 1-4-chloro-2-nitrophenyl pyrrolidine #, 1-4-chloranyl-2-nitro-phenyl pyrrolidine, 2-pyrrolidin-1-yl-5-chloro-1-nitrobenzene
285321
1-(4-chloro-2-nitrophenyl)pyrrolidine
C1CCN(C1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]
This item is not returnable. View return policy
41173-36-8
C10H11ClN2O2
226.66
QATZMIJXHKOELL-UHFFFAOYSA-N
1-4-chloro-2-nitrophenyl pyrrolidine, 1-4-chloro-2-nitro-phenyl pyrrolidine, n-4-chloro-2-nitrophenyl pyrrolidine, 4-chloro-2-nitrophenyl pyrrolidine, qatzmijxhkoell-uhfffaoysa, 1-4-chloro-2-nitro-phenyl-pyrrolidine, 1-4-chloro-2-nitrophenyl pyrrolidine #, 1-4-chloranyl-2-nitro-phenyl pyrrolidine, 2-pyrrolidin-1-yl-5-chloro-1-nitrobenzene
285321
1-(4-chloro-2-nitrophenyl)pyrrolidine
C1CCN(C1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 915044
Formula Weight 226.66
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(4-Chloro-2-nitrophenyl)pyrrolidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'