Läs mer
1-(4-Chloro-3-trifluoromethylphenyl)-3-(4-hydroxyphenyl)urea, TRC
Produktkod. 30358046
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30358046 100 mg
30362140 50 mg
30362982 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30358046 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC596195100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1129683-83-5
Molekylformel C14H10ClF3N2O2
Molekylvikt (g/mol) 330.69
Synonym N-[4-Chloro-3-(trifluoromethyl)phenyl]-N'-(4-hydroxyphenyl)urea
IUPAC-namn 1-[4-chloro-3-(trifluoromethyl)phenyl]-3-(4-hydroxyphenyl)urea
LEDER Oc1ccc(NC(=O)Nc2ccc(Cl)c(c2)C(F)(F)F)cc1
Specifikationer
Kemiskt namn eller material 1-(4-Chloro-3-trifluoromethylphenyl)-3-(4-hydroxyphenyl)urea
Kvantitet 100 mg
InChI formel InChI=1S/C14H10ClF3N2O2/c15-12-6-3-9(7-11(12)14(16,17)18)20-13(22)19-8-1-4-10(21)5-2-8/h1-7,21H,(H2,19,20,22)