Learn More
1-(4-Chlorophenyl)-1H-pyrrole-2-carbaldehyde, Tech., Thermo Scientific™
Catalog Number p-2694323
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
37560-50-2
C11H8ClNO
205.641
MFCD00174283
NNQOVJLQFYROBU-UHFFFAOYSA-N
1-4-chlorophenyl-1h-pyrrole-2-carbaldehyde, 1-4-chlorophenyl pyrrole-2-carbaldehyde, 1-p-chlorphenyl-2-pyrrolcarbaldehyd, 1-4-chloro-phenyl-1h-pyrrole-2-carbaldehy de, 1h-pyrrole-2-carboxaldehyde,1-4-chlorophenyl
737212
1-(4-chlorophenyl)pyrrole-2-carbaldehyde
C1=CN(C(=C1)C=O)C2=CC=C(C=C2)Cl
37560-50-2
C11H8ClNO
205.641
MFCD00174283
NNQOVJLQFYROBU-UHFFFAOYSA-N
1-4-chlorophenyl-1h-pyrrole-2-carbaldehyde, 1-4-chlorophenyl pyrrole-2-carbaldehyde, 1-p-chlorphenyl-2-pyrrolcarbaldehyd, 1-4-chloro-phenyl-1h-pyrrole-2-carbaldehy de, 1h-pyrrole-2-carboxaldehyde,1-4-chlorophenyl
737212
1-(4-chlorophenyl)pyrrole-2-carbaldehyde
C1=CN(C(=C1)C=O)C2=CC=C(C=C2)Cl
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 205.64
Percent Purity Tech.
Grade Technical
Chemical Name or Material 1-(4-Chlorophenyl)-1H-pyrrole-2-carbaldehyde