Learn More
1,4-Diacetylbenzene, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 1009-61-6 |
| Molecular Formula | C10H10O2 |
| MDL Number | MFCD00008750 |
| Quantity | 250 mg |
| SMILES | CC(C1=CC=C(C(C)=O)C=C1)=O |
| Molecular Weight (g/mol) | 162.19 |
| Percent Purity | 95% |
| Chemical Name or Material | 1,4-Diacetylbenzene |