missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dibromo-2,5-dimethoxybenzene, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
It is employed as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is employed as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 144°C to 148°C |
Quantity | 10 g |
Beilstein | 1953103 |
Solubility Information | Insoluble in water. |
Formula Weight | 295.96 |
Percent Purity | ≥98% |
Chemical Name or Material | 1,4-Dibromo-2,5-dimethoxybenzene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'