Learn More
1,4-Dibromotetrafluorobenzene, 99%, Thermo Scientific Chemicals
Catalog Number p-7024008
Click to view available options
Quantity:
1 g
5 g
25 g
344-03-6
C6Br2F4
307.87
MFCD00000309
QFTZULJNRAHOIY-UHFFFAOYSA-N
1,4-dibromotetrafluorobenzene, p-dibromotetrafluorobenzene, benzene, 1,4-dibromo-2,3,5,6-tetrafluoro, benzene, 1,4-dibromotetrafluoro, 1,3,5,6-tetrafluorobenzene, benzene,4-dibromotetrafluoro, emolecules 536967, tetrafluoro-1,4-dibromo benzene, perfluoro-1,4-dibromobenzene
67653
1,4-dibromo-2,3,5,6-tetrafluorobenzene
FC1=C(F)C(Br)=C(F)C(F)=C1Br
344-03-6
C6Br2F4
307.87
MFCD00000309
QFTZULJNRAHOIY-UHFFFAOYSA-N
1,4-dibromotetrafluorobenzene, p-dibromotetrafluorobenzene, benzene, 1,4-dibromo-2,3,5,6-tetrafluoro, benzene, 1,4-dibromotetrafluoro, 1,3,5,6-tetrafluorobenzene, benzene,4-dibromotetrafluoro, emolecules 536967, tetrafluoro-1,4-dibromo benzene, perfluoro-1,4-dibromobenzene
67653
1,4-dibromo-2,3,5,6-tetrafluorobenzene
FC1=C(F)C(Br)=C(F)C(F)=C1Br
It is used as a chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a chemical intermediate.
Solubility
Solubility in methanol is almost transparent.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point 78°C to 81°C
Quantity 1 g
Formula Weight 307.87
Percent Purity 99%
Chemical Name or Material 1,4-Dibromotetrafluorobenzene