Läs mer
1,4-Dichlorophthalazine, TRC
Produktkod. 30370126
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30370126 1 g
30369248 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30370126 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4361651G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,4-Dichlorophthalazine
CAS 4752-10-7
Molekylformel C8H4Cl2N2
Kvantitet 1 g
InChI formel InChI=1S/C8H4Cl2N2/c9-7-5-3-1-2-4-6(5)8(10)12-11-7/h1-4H
LEDER Clc1nnc(Cl)c2ccccc12
Molekylvikt (g/mol) 199.04
Formel vikt 197.9752