Läs mer
1,4-Dihydro-1-methyl-4-oxo-3-pyridinecarboxamide, TRC
Produktkod. 30011036
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011036 1 mg
30008167 10 mg
30008232 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011036 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4534151MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 769-49-3
Molekylformel C7 H8 N2 O2
Molekylvikt (g/mol) 152.15
Synonym 3-Pyridinecarboxamide, 1,4-dihydro-1-methyl-4-oxo-, Nicotinamide, 1,4-dihydro-1-methyl-4-oxo- (6CI,7CI,8CI), 1,4-Dihydro-1-methyl-4-oxo-3-pyridinecarboxamide, 1-Methyl-1,4-dihydro-4-oxo-3-pyridinecarboxamide, 1-Methyl-4-pyridone-3-carboxamide, 1-Methyl-4-pyridone-5-carboxamide, 3-(Aminocarbonyl)-1-methyl-4(1H)-pyridone, 5-Aminocarbonyl-1-methyl-4(1H)-pyridone, N1-Methyl-4-pyridone-3-carboxamide, N1-Methyl-4-pyridone-3-carboxamide, N1-Methyl-4-pyridone-5-carboxamide, N1-Methyl-4-pyridone-5-carboxamide
IUPAC-namn 1-methyl-4-oxopyridine-3-carboxamide
LEDER CN1C=CC(=O)C(=C1)C(=O)N
Specifikationer
Kemiskt namn eller material N-Methyl-4-pyridone-3-carboxamide
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C7H8N2O2/c1-9-3-2-6(10)5(4-9)7(8)11/h2-4H,1H3,(H2,8,11)
Formel vikt 152.0586
Procent renhet >95%