Läs mer
1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 3-Methyl Ester, TRC
Produktkod. 30136611
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136611 500 mg
30011668 250 mg
17997924 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136611 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD449123500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 74936-72-4
Molekylformel C16 H16 N2 O6
Molekylvikt (g/mol) 332.31
Synonym 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylic Acid Methyl Ester
IUPAC-namn 5-methoxycarbonyl-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3-carboxylic acid
LEDER COC(=O)C1=C(C)NC(=C(C1c2cccc(c2)[N+](=O)[O-])C(=O)O)C
Specifikationer
Kemiskt namn eller material 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylic Acid Methyl Ester
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C16H16N2O6/c1-8-12(15(19)20)14(13(9(2)17-8)16(21)24-3)10-5-4-6-11(7-10)18(22)23/h4-7,14,17H,1-3H3,(H,19,20)
Formel vikt 332.1008
Procent renhet >95%