Läs mer
1,4-Dioxacyclotetradecane-5,14-dione, TRC
Produktkod. 17938984
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17938984 5 mg
17927264 1 mg
17958504 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17938984 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD7755405MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5578-82-5
Molekylformel C12 H20 O4
Molekylvikt (g/mol) 228.28
Synonym Cyclic Ethylene Ester Sebacic Acid, Cyclic Ethylene Decanedioate, Cyclic 1,2-Ethanediyl Ester Decanedioic Acid, Cyclic Sebacate Ethylene Glycol, Ethylene Sebacate;
IUPAC-namn 1,4-dioxacyclotetradecane-5,14-dione
LEDER O=C1CCCCCCCCC(=O)OCCO1
Specifikationer
Kemiskt namn eller material 1,4-Dioxacyclotetradecane-5,14-dione
Kvantitet 5 mg
InChI formel InChI=1S/C12H20O4/c13-11-7-5-3-1-2-4-6-8-12(14)16-10-9-15-11/h1-10H2
Formel vikt 228.1362