Läs mer
1,4-Dioxane-13C4, TRC
Produktkod. 30370384
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30370384 5 mg
1 options
Produktkod. 30370384 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4853575MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,4-Dioxane-13C4
CAS 1228182-37-3
Molekylformel 13C4H8O2
Kvantitet 5 mg
Synonym p-Dioxane-13C4 (8CI), 1,4-Diethylene-13C4 Dioxide, 1,4-Dioxacyclohexane-13C4, 1,4-Dioxan-13C4, Tetrahydro-1,4-dioxin-13C4, Diethylene Dioxide-13C, Diethylene Ether-13C, Diethylene Oxide-13C, Dioxan-13C, Dioxane-13C, Dioxyethylene-13C Ether, NE 220-13C, NSC 8728-13C, p-Dioxan-13C
InChI formel InChI=1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2/i1+1,2+1,3+1,4+1
LEDER [13CH2]1O[13CH2][13CH2]O[13CH2]1
Molekylvikt (g/mol) 92.08