missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Fluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H62721.06

Additional Details : CAS Number : 23788-28-5 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15477776

  • 11319.11 SEK / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Fluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea
298.24
3066601
23788-28-5
MFCD00418715

Specifications

1-(4-Fluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea
23788-28-5
97%
MFCD00418715
5g
Solid
298.24
3066601
Videos
SDS