missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Iodophenyl)pyrrole, 97%, Thermo Scientific Chemicals

Catalog Number p-7022491
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
92636-36-7
C10H8IN
269.085
MFCD00052399
FMURNAZHVQDQQN-UHFFFAOYSA-N
1-4-iodophenyl pyrrole, 1-4-iodophenyl-1h-pyrrole, acmc-20apaj, 4-iodophenyl pyrrole, maybridge1_002926, 1-4-iodo-phenyl-1h-pyrrole, 1h-pyrrole,1-4-iodophenyl, 1-4-iodophenyl-1h-pyrrole #
272430
1-(4-iodophenyl)pyrrole
C1=CN(C=C1)C2=CC=C(C=C2)I
This item is not returnable. View return policy
92636-36-7
C10H8IN
269.085
MFCD00052399
FMURNAZHVQDQQN-UHFFFAOYSA-N
1-4-iodophenyl pyrrole, 1-4-iodophenyl-1h-pyrrole, acmc-20apaj, 4-iodophenyl pyrrole, maybridge1_002926, 1-4-iodo-phenyl-1h-pyrrole, 1h-pyrrole,1-4-iodophenyl, 1-4-iodophenyl-1h-pyrrole #
272430
1-(4-iodophenyl)pyrrole
C1=CN(C=C1)C2=CC=C(C=C2)I

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 128°C to 133°C
Quantity 1 g
Beilstein 7915717
Sensitivity Light sensitive
Formula Weight 269.09
Percent Purity 97%
Chemical Name or Material 1-(4-Iodophenyl)pyrrole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'