Learn More
1-(4-Methoxyphenyl)acetophenone, TRC
Product Code. 30470265
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30470265 100 mg 100mg
1 options
Product Code. 30470265 Supplier Toronto Research Chemicals Supplier No. TRCM265800100MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(4-Methoxyphenyl)acetophenone
CAS 1023-17-2
Molecular Formula C15H14O2
Quantity 100 mg
Synonym 1-(4-Methoxyphenyl)-2-phenyl-ethanone, 2-Phenyl-p-methoxyacetophenone, 4-Methoxy-alpha-phenylacetophenone, 4-Methoxydeoxybenzoin, Benzyl p-Methoxyphenyl Ketone, NSC 26658, p-Anisyl Benzyl Ketone
InChI Formula InChI=1S/C15H14O2/c1-17-14-9-7-13(8-10-14)15(16)11-12-5-3-2-4-6-12/h2-10H,11H2,1H3
SMILES COc1ccc(cc1)C(=O)Cc2ccccc2
IUPAC Name 1-(4-methoxyphenyl)-2-phenylethanone
Molecular Weight (g/mol) 226.27
Formula Weight 226.0994