Läs mer
1-(4-Methylphenyl)-1-methylethylamine Hydrochloride, TRC
Produktkod. 30473605
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30473605 50 mg
30478566 500 mg
30468121 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30473605 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33929550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1654774-17-0
Molekylformel C10H15N . HCl
Molekylvikt (g/mol) 185.69
IUPAC-namn 2-(4-methylphenyl)propan-2-amine;hydrochloride
LEDER Cl.Cc1ccc(cc1)C(C)(C)N
Specifikationer
Kemiskt namn eller material 1-(4-Methylphenyl)-1-methylethylamine Hydrochloride
Kvantitet 50 mg
InChI formel InChI=1S/C10H15N.ClH/c1-8-4-6-9(7-5-8)10(2,3)11;/h4-7H,11H2,1-3H3;1H
Formel vikt 185.0971