Läs mer
1-(4-Nitrobenzyl)-1H-1,2,4-triazole, TRC
Produktkod. 30180014
Change view
Kvantitet:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180014 500 mg
30179732 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30180014 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN900988500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 119192-09-5
Molekylformel C9H8N4O2
Molekylvikt (g/mol) 204.18
Synonym 1-[(4-Nitrophenyl)methyl]-1H-1,2,4-triazole, 1-(4-Nitrobenzyl)-1,2,4-triazole, 1-(4-Nitrophenyl)methyl-1,2,4-triazole, 1-[(4-Nitrophenyl)methyl]-1,2,4-triazole, 4-(1,2,4-Triazol-1-ylmethyl)nitrobenzene;
IUPAC-namn 1-[(4-nitrophenyl)methyl]-1,2,4-triazole
LEDER [O-][N+](=O)c1ccc(Cn2cncn2)cc1
Specifikationer
Kemiskt namn eller material 1-(4-Nitrobenzyl)-1H-1,2,4-triazole
Kvantitet 500 mg
InChI formel InChI=1S/C9H8N4O2/c14-13(15)9-3-1-8(2-4-9)5-12-7-10-6-11-12/h1-4,6-7H,5H2
Formel vikt 204.0647