missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Nitrophenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-7029857
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
142818-03-9
C11H6F3N3O4
301.18
MFCD00173867
FGHNILTUXGDLKS-UHFFFAOYSA-N
1-4-nitrophenyl-5-trifluoromethyl pyrazole-4-carboxylic acid, 1-4-nitrophenyl-5-trifluoromethyl-1h-pyrazole-4-carboxylic acid, 1-4-nitrophenyl-5-trifluoromethyl-1h-pyrazole, maybridge1_003970, acmc-20apa6, 5-trifluoromethyl-1-4-nitrophenyl-1h-pyrazole-4-carboxylic acid
688692
1-(4-nitrophenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
OC(=O)C1=C(N(N=C1)C1=CC=C(C=C1)[N+]([O-])=O)C(F)(F)F
Este artículo no se puede devolver. Vea la política de devoluciones
142818-03-9
C11H6F3N3O4
301.18
MFCD00173867
FGHNILTUXGDLKS-UHFFFAOYSA-N
1-4-nitrophenyl-5-trifluoromethyl pyrazole-4-carboxylic acid, 1-4-nitrophenyl-5-trifluoromethyl-1h-pyrazole-4-carboxylic acid, 1-4-nitrophenyl-5-trifluoromethyl-1h-pyrazole, maybridge1_003970, acmc-20apa6, 5-trifluoromethyl-1-4-nitrophenyl-1h-pyrazole-4-carboxylic acid
688692
1-(4-nitrophenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
OC(=O)C1=C(N(N=C1)C1=CC=C(C=C1)[N+]([O-])=O)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 200°C to 204°C
Quantity 1 g
Formula Weight 301.19
Percent Purity 97%
Chemical Name or Material 1-(4-Nitrophenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'