Läs mer
1,4-Phenylenediacetic Acid Ethyl Ester, TRC
Produktkod. 30421251
Change view
Kvantitet:
2.5 g
Förpackningsstorlek:
2.5g
Produktkod. Kvantitet unitSize
30421251 2.5 g 2.5g
1 options
Produktkod. 30421251 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3197302.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,4-Phenylenediacetic Acid Ethyl Ester
CAS 113520-36-8
Molekylformel C12H14O4
Kvantitet 2.5 g
Synonym 1,4-Benzenediacetic Acid Ethyl Ester, p-Benzenediacetic Acid Ethyl Ester, p-Phenylenediacetic Acid Ethyl Ester
InChI formel InChI=1S/C12H14O4/c1-2-16-12(15)8-10-5-3-9(4-6-10)7-11(13)14/h3-6H,2,7-8H2,1H3,(H,13,14)/p-1
LEDER CCOC(=O)CC1=CC=C(C=C1)CC(=O)[O-]
Molekylvikt (g/mol) 222.24