Läs mer
1-(4-Piperidinyl)ethanone Hydrochloride, TRC
Produktkod. 30411367
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30411367 1 g
1 options
Produktkod. 30411367 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP4820301G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-(4-Piperidinyl)ethanone Hydrochloride
CAS 89895-06-7
Molekylformel C7H14ClNO
Kvantitet 1 g
Synonym 1-(Piperidin-4-yl)ethanone Hydrochloride, 4-Acetylpiperidine Hydrochloride, Methyl 4-Piperidyl Ketone Hydrochloride
InChI formel InChI=1S/C7H13NO.ClH/c1-6(9)7-2-4-8-5-3-7;/h7-8H,2-5H2,1H3;1H
LEDER O=C(C)C1CCNCC1.Cl
Molekylvikt (g/mol) 163.65