Läs mer
1-[(4R,5R)-N2-(10,12-Dimethyl-1-oxotetradecyl)-4-hydroxy-5-(phenylthio)-L-ornithine]-pneumocandin B0, TRC
Produktkod. 30179942
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179942 25 mg
30180212 10 mg
30180386 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179942 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD46575025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 198706-35-3
Specifikationer
Formatera Neat
Molekylformel C56H84N8O16S
Molekylvikt (g/mol) 1157.38, 1157.39
InChI-nyckel SXBBAHGGUQHBIH-LUBNTDDFSA-N
LEDER [H][C@@]12C[C@@H](O)CN1C(=O)[C@@]([H])(NC(=O)[C@H](C[C@@H](O)[C@H](NC(=O)[C@]1([H])[C@@H](O)CCN1C(=O)[C@@]([H])(NC(=O)[C@@]([H])(NC2=O)[C@H](O)[C@@H](O)C1=CC=C(O)C=C1)[C@H](O)CC(N)=O)SC1=CC=CC=C1)NC(=O)CCCCCCCCC(C)CC(C)CC)[C@@H](C)O
IUPAC-namn N-[(3S,6S,9S,11R,15S,18S,20R,21R,24S,25S)-3-[(1R)-2-carbamoyl-1-hydroxyethyl]-6-[(1S,2S)-1,2-dihydroxy-2-(4-hydroxyphenyl)ethyl]-11,20,25-trihydroxy-15-[(1R)-1-hydroxyethyl]-2,5,8,14,17,23-hexaoxo-21-(phenylsulfanyl)-1,4,7,13,16,22-hexaazatricyclo[22.3.0.0⁹,¹³]heptacosan-18-yl]-10,12-dimethyltetradecanamide