Läs mer
1-[(4R,5R)-N2-(10,12-Dimethyl-1-oxotetradecyl)-4-hydroxy-5-(phenylthio)-L-ornithine]-pneumocandin B0, TRC
Produktkod. 30180386
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180386 5 mg
30179942 25 mg
30180212 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30180386 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4657505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 198706-35-3
Specifikationer
Formatera Neat
Molekylvikt (g/mol) 1157.38