missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
1-[5-Amino-2-[(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]ethanone Dihydrochloride, Mikromol™
Description
1-[5-Amino-2-[(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]ethanone Dihydrochloride
Specifications
Specifications
| Kemiskt namn eller material | 1-[5-Amino-2-[(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]ethanone Dihydrochloride |
| CAS | 2732673-51-5 |
| Analyt- eller komponentnamn | 1-[5-Amino-2-[(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]ethanone Dihydrochloride |
| Frakt skick | Room Temperature |
| Molekylformel | C14 H22 N2 O3 . 2 Cl H |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | Acebutolol Hydrochloride Imp. D (EP), Acebutolol Imp. D (EP), 1-[5-Amino-2-[(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]ethanone Dihydrochloride, Acebutolol Hydrochloride Imp. D (EP) as Dihydrochloride, Acebutolol Hydrochloride Impurity D as Dihydrochloride, Acebutolol Impurity D as Dihydrochloride |
| InChI formel | InChI=1S/C47H53ClN3O3.BrH/c1-49(2)43(52)46(38-17-9-5-10-18-38,39-19-11-6-12-20-39)31-35-51(33-29-45(54,30-34-51)37-25-27-42(48)28-26-37)36-32-47(44(53)50(3)4,40-21-13-7-14-22-40)41-23-15-8-16-24-41;/h5-28,54H,29-36H2,1-4H3;1H/q+1;/p-2 |
| LEDER | Cl.Cl.CC(C)NCC(O)COc1ccc(N)cc1C(=O)C |
| IUPAC-namn | 1-[5-amino-2-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]ethanone;dihydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?