Läs mer
1,5-Bis-(4-Chlorophenyl)biguanide Hydrochloride, Mikromol™
Beskrivning
1,5-Bis-(4-Chlorophenyl)biguanide Hydrochloride
Specifikationer
Specifikationer
| Kemiskt namn eller material | 1,5-bis(4-chlorophenyl)biguanide hydrochloride |
| CAS | 13590-98-2 |
| Analyt- eller komponentnamn | 1,5-bis(4-chlorophenyl)biguanide hydrochloride |
| Frakt skick | Room Temperature |
| Molekylformel | C14 H13 Cl2 N5 . Cl H |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | N,N'-bis(4-Chlorophenyl)imidodicarbonimidic Diamide Hydrochloride (1:1), Biguanide, 1,5-bis(p-chlorophenyl)-, monohydrochloride (8CI), Imidodicarbonimidic diamide, N,N'-bis(4-chlorophenyl)-, monohydrochloride (9CI), ST 39008, 3583W95, Ph Eur Proguanil Impurity C, USP Proguanil Related Compound C, CCI 14231, Proguanil Related Compound C as Hydrochloride, Proguanil Hydrochloride Impurity C as Hydrochloride, Proguanil Impurity C as Hydrochloride |
| InChI formel | InChI=1S/C14H13Cl2N5.ClH/c15-9-1-5-11(6-2-9)19-13(17)21-14(18)20-12-7-3-10(16)4-8-12;/h1-8H,(H5,17,18,19,20,21);1H |
| LEDER | Cl.Clc1ccc(NC(=N)NC(=N)Nc2ccc(Cl)cc2)cc1 |
| IUPAC-namn | 1-(4-chlorophenyl)-3-[N-(4-chlorophenyl)carbamimidoyl]guanidine;hydrochloride |
| Visa mer |