Läs mer
1,5-Dihydroxyanthraquinone, tech. 90%, Thermo Scientific™
Produktkod. 11304324
Kvantitet:
10 g
100 g
500 g
Förpackningsstorlek:
100g
10g
500g
Produktkod. 11304324
Brand: Thermo Scientific Alfa Aesar L00848.09
Request Bulk or Custom Format
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 117-12-4
Molekylformel C14H8O4
Molekylvikt (g/mol) 240.21
MDL-nummer MFCD00001210
InChI-nyckel JPICKYUTICNNNJ-UHFFFAOYSA-N
Synonym anthrarufin, 1,5-dihydroxyanthraquinone, 1,5-dihydroxyanthrachinon, 9,10-anthracenedione, 1,5-dihydroxy, 1,5-dihydroxy-9,10-anthraquinone, 1,5-dihydroxy-9,10-anthracenedione, anthraquinone, 1,5-dihydroxy, az-u, unii-kpb60w5s3m, ccris 3150
PubChem CID 8328
ChEBI CHEBI:37501
IUPAC-namn 1,5-dihydroxyanthracene-9,10-dione
LEDER OC1=CC=CC2=C1C(=O)C1=CC=CC(O)=C1C2=O
Specifikationer
Kemiskt namn eller material 1,5-Dihydroxyanthraquinone
Smältpunkt ∽280°C (decomposition)
Kvantitet 10 g
Beilstein 1881718
Merck Index 14,689
Formel vikt 240.21
Procent renhet 90%
Kvalitet Technical