Läs mer
1,5-Diisocyanatonaphthalene, TRC
Produktkod. 30041152
Change view
Kvantitet:
250 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30041152 250 mg
30040365 500 mg
30012600 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30041152 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD679083250MG
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3173-72-6
Molekylformel C12H6N2O2
Molekylvikt (g/mol) 210.19
Synonym 1,5-Diisocyanatonaphthalene (ACI), 1,5-Naphthalenediol, diisocyanate (5CI), Isocyanic acid, 1,5-naphthylene ester (6CI, 8CI), 1,5-Naphthalene diisocyanate, 1,5-Naphthyl diisocyanate, 1,5-Naphthylene diisocyanate, 1,5-NDI, Naphthalene-1,5-diyl diisocyanate, NDI, NSC 240728
LEDER C1=CC2=C(C=CC=C2N=C=O)C(=C1)N=C=O
Specifikationer
Kemiskt namn eller material 1,5-Diisocyanatonaphthalene
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C12H6N2O2/c15-7-13-11-5-1-3-9-10(11)4-2-6-12(9)14-8-16/h1-6H
Formel vikt 210.0429
Procent renhet >95%