missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,5-Dimethyl-1H-pyrazole-3-carboxylic acid, 98%

Brand:  Alfa Aesar™ H64475.06

Additional Details : CAS Number : 5744-59-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15451699

  • 1613.01 SEK / 5g


Estimated Shipment date
from Supplier 23-01-2019
Add to basket

Specifications

Chemical Identifiers

5744-59-2
140.142
PXRXGHUTKHXUGF-UHFFFAOYSA-N
587757
CC1=CC(=NN1C)C(=O)O
C6H8N2O2
MFCD00085040
1,5-dimethyl-1h-pyrazole-3-carboxylic acid, 1,5-dimethyl-1h-pyrazole-3-carboxylicacid, 1,5-dimethyl-3-pyrazolecarboxylic acid, 1h-pyrazole-3-carboxylic acid, 1,5-dimethyl, 1,5-dimethyl-1h-pyrazole-3-carboxylic, acmc-209lyb, pyrazole-3-carboxylic acid, 1,5-dimethyl, 1h-pyrazole-3-carboxylicacid, 1,5-dimethyl, 1,5-dimethyl-1h-pyrazole-3-carboxylic acid #, 1,5-dimethyl-1h-pyrazole-3-carboxylic acid
1,5-dimethylpyrazole-3-carboxylic acid

Specifications

1,5-Dimethyl-1H-pyrazole-3-carboxylic acid
5g
C6H8N2O2
115917
PXRXGHUTKHXUGF-UHFFFAOYSA-N
1,5-dimethylpyrazole-3-carboxylic acid
587757
Solid
170°C to 174°C
5744-59-2
MFCD00085040
1,5-dimethyl-1h-pyrazole-3-carboxylic acid, 1,5-dimethyl-1h-pyrazole-3-carboxylicacid, 1,5-dimethyl-3-pyrazolecarboxylic acid, 1h-pyrazole-3-carboxylic acid, 1,5-dimethyl, 1,5-dimethyl-1h-pyrazole-3-carboxylic, acmc-209lyb, pyrazole-3-carboxylic acid, 1,5-dimethyl, 1h-pyrazole-3-carboxylicacid, 1,5-dimethyl, 1,5-dimethyl-1h-pyrazole-3-carboxylic acid #, 1,5-dimethyl-1h-pyrazole-3-carboxylic acid
CC1=CC(=NN1C)C(=O)O
140.142
140.14
98%
Videos
SDS