Läs mer
1-(5-O-Acetyl-beta-D-ribofuranosyl)-1H-1,2,4-triazole-3-carboxamide (5'-O-Acetylribavirin), Mikromol™
Produktkod. 30458201
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30458201 4 x 25 mg
30052646 25 mg
18040580 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30458201 Leverantör Mikromol Leverantörsnummer MM0542.0600254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(5-O-Acetyl-beta-D-ribofuranosyl)-1H-1,2,4-triazole-3-carboxamide (5'-O-Acetylribavirin), Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-(5-O-Acetyl-beta-D-ribofuranosyl)-1H-1,2,4-triazole-3-carboxamide (5'-O-Acetylribavirin)
CAS 58151-87-4
Analyt- eller komponentnamn 1-(5-O-Acetyl-beta-D-ribofuranosyl)-1H-1,2,4-triazole-3-carboxamide (5'-O-Acetylribavirin)
Frakt skick Room Temperature
Molekylformel C10 H14 N4 O6
Certifikat och efterlevnad ISO 17025
Synonym 1-(5-O-Acetyl-beta-D-ribofuranosyl)-1H-1,2,4-triazole-3-carboxamide, Ribavirin Imp. F (EP), 5'-O-Acetylribavirin, Ribavirin Impurity F
InChI formel InChI=1S/C10H14N4O6/c1-4(15)19-2-5-6(16)7(17)10(20-5)14-3-12-9(13-14)8(11)18/h3,5-7,10,16-17H,2H2,1H3,(H2,11,18)/t5-,6-,7-,10-/m1/s1
Lösningstyp Neat
LEDER CC(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc(n2)C(=O)N
IUPAC-namn [(2R,3S,4R,5R)-5-(3-carbamoyl-1,2,4-triazol-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl acetate
Molekylvikt (g/mol) 286.24
Kvantitet 4 x 25 mg
Formel vikt 286.0913 g/mol