Läs mer
1,5-Pentanediyl Bismethanethiosulfonate, TRC
Produktkod. 30419505
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30419505 25 mg
30409839 10 mg
30415043 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30419505 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP27325025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56-00-8
Molekylformel C7 H16 O4 S4
Molekylvikt (g/mol) 292.46
IUPAC-namn 1,5-bis(methylsulfonylsulfanyl)pentane
LEDER CS(=O)(=O)SCCCCCSS(=O)(=O)C
Specifikationer
Kemiskt namn eller material 1,5-Pentanediyl Bismethanethiosulfonate
Kvantitet 25 mg
InChI formel InChI=1S/C7H16O4S4/c1-14(8,9)12-6-4-3-5-7-13-15(2,10)11/h3-7H2,1-2H3
Formel vikt 291.9931