Läs mer
1,6,13,18-Tetraoxacyclotetracosane-7,12,19,24-tetrone, TRC
Produktkod. 30451158
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30451158 50 mg
30452940 25 mg
30452350 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30451158 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT30601050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 78837-87-3
Molekylformel C20H32O8
Molekylvikt (g/mol) 400.46
Synonym 1,6,13,18-Tetraoxacyclotetracosane-7,12,19,24-tetraone
IUPAC-namn 1,6,13,18-tetraoxacyclotetracosane-7,12,19,24-tetrone
LEDER O=C1CCCCC(=O)OCCCCOC(=O)CCCCC(=O)OCCCCO1
Specifikationer
Kemiskt namn eller material 1,6,13,18-Tetraoxacyclotetracosane-7,12,19,24-tetrone
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C20H32O8/c21-17-9-1-2-10-18(22)26-14-7-8-16-28-20(24)12-4-3-11-19(23)27-15-6-5-13-25-17/h1-16H2
Formel vikt 400.2097
Procent renhet >95%