Läs mer
1,6,7,12-Tetrachloroperylene Tetracarboxylic Acid Dianhydride, TRC
Produktkod. 30453656
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30453656 500 mg
1 options
Produktkod. 30453656 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT889865500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,6,7,12-Tetrachloroperylene Tetracarboxylic Acid Dianhydride
CAS 156028-26-1
Molekylformel C24H4Cl4O6
Kvantitet 500 mg
Synonym 5,6,12,13-tetrachloro-Perylo[3,4-cd:9,10-c'd']dipyran-1,3,8,10-tetrone, 5,6,12,13-Tetrachloroperylo[3,4-cd:9,10-c'd']dipyran-1,3,8,10-tetrone, 1,6,7,12-Tetrachloro-3,4:9,10-perylenetetracarboxylic dianhydride, 1,6,7,12-Tetrachloroperylene-3,4:9,10-tetracarboxylic acid dianhydride, 1,6,7,12-Tetrachloroperylene-3,4:9,10-tetracarboxylic dianhydride, 5,6,12,13-Tetrachloroperylenetetracarboxylic dianhydride
InChI formel InChI=1S/C24H4Cl4O6/c25-9-1-5-13-6(22(30)33-21(5)29)2-11(27)17-18-12(28)4-8-14-7(23(31)34-24(8)32)3-10(26)16(20(14)18)15(9)19(13)17/h1-4H
LEDER C1=C2C3=C4C(=C1Cl)C5=C(C=C6C7=C5C(=C(C=C7C(=O)OC6=O)Cl)C4=C(C=C3C(=O)OC2=O)Cl)Cl
Molekylvikt (g/mol) 530.1
Formel vikt 527.8762