Läs mer
1-(6-Amino-9-Beta-D-ribofuranosyl-9H-purin-2-yl)-1H-pyrazole-4-carboxylic AcidEthyl Ester, TRC
Produktkod. 30325968
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30325968 50 mg
1 options
Produktkod. 30325968 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA62921050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-(6-Amino-9-Beta-D-ribofuranosyl-9H-purin-2-yl)-1H-pyrazole-4-carboxylic AcidEthyl Ester
CAS 313348-16-2
Molekylformel C16H19N7O6
Kvantitet 50 mg
Synonym CVT 3127, Regadenoson Impurity
InChI formel InChI=1S/C16H19N7O6/c1-2-28-15(27)7-3-19-23(4-7)16-20-12(17)9-13(21-16)22(6-18-9)14-11(26)10(25)8(5-24)29-14/h3-4,6,8,10-11,14,24-26H,2,5H2,1H3,(H2,17,20,21)/t8-,10-,11-,14-/m1/s1
LEDER CCOC(=O)c1cnn(c1)c2nc(N)c3ncn([C@@H]4O[C@H](CO)[C@@H](O)[C@H]4O)c3n2
IUPAC-namn ethyl 1-[6-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]purin-2-yl]pyrazole-4-carboxylate
Molekylvikt (g/mol) 405.37