Läs mer
1-(6-Amino-9H-purin-2-yl)-1H-pyrazole-4-carboxylic Acid Ethyl Ester (>85%), TRC
Produktkod. 30180269
Change view
Kvantitet:
5 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180269 250 mg
30180253 5 mg
30180132 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30180269 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA919953250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1702334-22-2
Molekylformel C11H11N7O2
Molekylvikt (g/mol) 273.25
Synonym Ethyl 1-(6-Amino-9H-purin-2-yl)-1H-pyrazole-4-carboxylate
IUPAC-namn ethyl 1-(6-amino-9H-purin-2-yl)pyrazole-4-carboxylate
LEDER CCOC(=O)c1cnn(c1)c2nc(N)c3nc[nH]c3n2
Specifikationer
Kemiskt namn eller material 1-(6-Amino-9H-purin-2-yl)-1H-pyrazole-4-carboxylic Acid Ethyl Ester
Kvantitet 250 mg
InChI formel InChI=1S/C11H11N7O2/c1-2-20-10(19)6-3-15-18(4-6)11-16-8(12)7-9(17-11)14-5-13-7/h3-5H,2H2,1H3,(H3,12,13,14,16,17)
Formel vikt 273.0974