Läs mer
1,6-Di-O-acetyl-2,3,4-tri-O-benzyl-Alpha-D-mannopyranose, TRC
Produktkod. 30368666
Change view
Kvantitet:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368666 1 g
30354506 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30368666 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4140001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,6-Di-O-acetyl-2,3,4-tri-O-benzyl-a-D-mannopyranose
CAS 65556-30-1
Molekylformel C31 H34 O8
Kvantitet 1 g
InChI formel InChI=1S/C31H34O8/c1-22(32)34-21-27-28(35-18-24-12-6-3-7-13-24)29(36-19-25-14-8-4-9-15-25)30(31(39-27)38-23(2)33)37-20-26-16-10-5-11-17-26/h3-17,27-31H,18-21H2,1-2H3/t27-,28-,29+,30+,31+/m1/s1
LEDER CC(=O)OC[C@H]1O[C@H](OC(=O)C)[C@@H](OCc2ccccc2)[C@@H](OCc3ccccc3)[C@@H]1OCc4ccccc4
IUPAC-namn [(2R,3R,4S,5S,6R)-6-acetyloxy-3,4,5-tris(phenylmethoxy)oxan-2-yl]methyl acetate
Molekylvikt (g/mol) 534.6
Formel vikt 534.2254