Läs mer
1,6-Dimethyl-4-(methylamino)pyridin-2(1H)-one, TRC
Produktkod. 30364545
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30364545 10 mg
30358373 5 mg
30354585 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30364545 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD15966110MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1934421-57-4
Molekylformel C8H12N2O
Molekylvikt (g/mol) 152.19
Synonym 1,6-Dimethyl-4-(methylamino)-1,2-dihydropyridin-2-one, 1,6-Dimethyl-4-(methylamino)pyridin-2-one
IUPAC-namn 1,6-dimethyl-4-(methylamino)pyridin-2-one
LEDER CNC1=CC(=O)N(C)C(=C1)C
Specifikationer
Kemiskt namn eller material 1,6-Dimethyl-4-(methylamino)pyridin-2(1H)-one
Kvantitet 10 mg
InChI formel InChI=1S/C8H12N2O/c1-6-4-7(9-2)5-8(11)10(6)3/h4-5,9H,1-3H3
Formel vikt 152.095