Läs mer
1,6-Dioxacyclododecane-7,12-dione, TRC
Produktkod. 30004557
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004557 1 mg
30003443 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30004557 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4853051MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 777-95-7
Molekylformel C10H16O4
Molekylvikt (g/mol) 200.23
Synonym Adipic Acid Cyclic Tetramethylene Ester
LEDER O=C1OCCCCOC(CCCC1)=O
Specifikationer
Kemiskt namn eller material 1,6-Dioxacyclododecane-7,12-dione
Anteckningar om renhetsgrad GC
Kvantitet 1 mg
InChI formel InChI=1S/C10H16O4/c11-9-5-1-2-6-10(12)14-8-4-3-7-13-9/h1-8H2
Formel vikt 200.1049
Procent renhet >95%