missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanedithiol, 97%, Thermo Scientific Chemicals

Catalog Number p-142388
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1191-43-1
C6H14S2
150.30
MFCD00004910
SRZXCOWFGPICGA-UHFFFAOYSA-N
1,6-hexanedithiol, 1,6-dimercaptohexane, 1,6-hexanedimercaptan, usaf uctl-72, hexamethylene dimercaptan, 1,6-hexane dithiol, 1,6-hexamethylenedithiol, unii-rwn7rm884e, fema no. 3495, hexanedithiol-1,6
14491
hexane-1,6-dithiol
SCCCCCCS
This item is not returnable. View return policy
1191-43-1
C6H14S2
150.30
MFCD00004910
SRZXCOWFGPICGA-UHFFFAOYSA-N
1,6-hexanedithiol, 1,6-dimercaptohexane, 1,6-hexanedimercaptan, usaf uctl-72, hexamethylene dimercaptan, 1,6-hexane dithiol, 1,6-hexamethylenedithiol, unii-rwn7rm884e, fema no. 3495, hexanedithiol-1,6
14491
hexane-1,6-dithiol
SCCCCCCS

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point -21.0°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 237.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula HS(CH2)6SH
Refractive Index 1.5110 to 1.5150
Quantity 5 g
Beilstein 01, 484
Specific Gravity 0.99
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in fat
Formula Weight 150.3
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 6-Hexanedithiol, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'