Läs mer
1-(6-Methyl-3-pyridinyl)-2-[4-(methylsulfonyl)phenyl]ethanone, Mikromol™
Produktkod. 17910946
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17910946 25 mg
17911136 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17910946 Leverantör Mikromol Leverantörsnummer MM0965.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(6-Methyl-3-pyridinyl)-2-[4-(methylsulfonyl)phenyl]ethanone
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-(6-Methyl-3-pyridinyl)-2-[4-(methylsulfonyl)phenyl]ethanone
CAS 221615-75-4
Analyt- eller komponentnamn 1-(6-Methyl-3-pyridinyl)-2-[4-(methylsulfonyl)phenyl]ethanone
Frakt skick Room Temperature
Molekylformel C15 H15 N O3 S
Certifikat och efterlevnad ISO 9001
Synonym 1-(6-Methyl-3-pyridinyl)-2-[4-(methylsulfonyl)phenyl]-ethanone, 1-(6-Methylpyridin-3-yl)-2-[4-(methylsulfonyl)phenyl]ethanone
InChI formel InChI=1S/C15H15NO3S/c1-11-3-6-13(10-16-11)15(17)9-12-4-7-14(8-5-12)20(2,18)19/h3-8,10H,9H2,1-2H3
LEDER Cc1ccc(cn1)C(=O)Cc2ccc(cc2)S(=O)(=O)C
IUPAC-namn 1-(6-methylpyridin-3-yl)-2-(4-methylsulfonylphenyl)ethanone
Molekylvikt (g/mol) 289.35
Kvantitet 25 mg
Formel vikt 289.0773 g/mol