Läs mer
1,7-Dimethyluric Acid, TRC
Produktkod. 30379421
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30379421 25 mg
30377308 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30379421 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD47969525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,7-Dimethyluric Acid
CAS 33868-03-0
Anteckningar om renhetsgrad HPLC
Molekylformel C7 H8 N4 O3
Kvantitet 25 mg
InChI formel InChI=1S/C7H8N4O3/c1-10-3-4(8-6(10)13)9-7(14)11(2)5(3)12/h1-2H3,(H,8,13)(H,9,14)
LEDER CN1C(=O)NC2=C(N(C)C(=O)N2)C1=O
IUPAC-namn 1,7-dimethyl-3,9-dihydropurine-2,6,8-trione
Molekylvikt (g/mol) 196.16
Formel vikt 196.0596