Läs mer
1,8-Diaminonaphthalene, TRC
Produktkod. 30357353
Change view
Kvantitet:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30357353 100 g
30351645 250 g
30353383 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30357353 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD416445100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 479-27-6
Molekylformel C10 H10 N2
Molekylvikt (g/mol) 158.2
Synonym 1,8-Diamine naphthaline, 1,8-Diaminonaphthalene, 1,8-Naphthylenediamine, NSC 6081
IUPAC-namn naphthalene-1,8-diamine
LEDER Nc1cccc2cccc(N)c12
Specifikationer
Kemiskt namn eller material 1,8-Diaminonaphthalene
Kvantitet 100 g
InChI formel InChI=1S/C10H10N2/c11-8-5-1-3-7-4-2-6-9(12)10(7)8/h1-6H,11-12H2
Formel vikt 158.0844