Läs mer
1,8-Diethyl-1,3,4,9-tetrahydro-pyrano[3,4-b]indole-1-acetic Acid Methyl Ester, TRC
Produktkod. 30369726
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30369726 50 mg
1 options
Produktkod. 30369726 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44503550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 2-[(1RS)-1,8-Diethyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetate (Etodolac Methyl Ester)
CAS 122188-02-7
Molekylformel C18 H23 N O3
Kvantitet 50 mg
Synonym Methyl 2-[(1RS)-1,8-Diethyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetate, Etodolac Methyl Ester, Pyrano[3,4-b]indole-1-acetic acid, 1,8-diethyl-1,3,4,9-tetrahydro-, methyl ester, Methyl 1,8-diethyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetate
InChI formel InChI=1S/C18H23NO3/c1-4-12-7-6-8-13-14-9-10-22-18(5-2,11-15(20)21-3)17(14)19-16(12)13/h6-8,19H,4-5,9-11H2,1-3H3
LEDER CCc1cccc2c3CCOC(CC)(CC(=O)OC)c3[nH]c12
IUPAC-namn methyl 2-(1,8-diethyl-4,9-dihydro-3H-pyrano[3,4-b]indol-1-yl)acetate
Molekylvikt (g/mol) 301.38
Formel vikt 301.1678