Läs mer
1-(9-Mercaptononyl)-3,6,9-trioxaundecan-11-ol, TRC
Produktkod. 30466918
Change view
Kvantitet:
5 mg
10 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30466918 5 mg
30465671 10 mg
30467411 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30466918 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2543255MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 130727-41-2
Molekylformel C17 H36 O4 S
Molekylvikt (g/mol) 336.53
IUPAC-namn 2-[2-[2-(11-sulfanylundecoxy)ethoxy]ethoxy]ethanol
LEDER OCCOCCOCCOCCCCCCCCCCCS
Specifikationer
Kemiskt namn eller material 1-(9-Mercaptononyl)-3,6,9-trioxaundecan-11-ol
Kvantitet 5 mg
InChI formel InChI=1S/C17H36O4S/c18-10-12-20-14-16-21-15-13-19-11-8-6-4-2-1-3-5-7-9-17-22/h18,22H,1-17H2
Formel vikt 336.2334