Läs mer
1-Acenaphthenol, TRC
Produktkod. 30012470
Change view
Kvantitet:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012470 1 g
30007226 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012470 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1307601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6306-07-6
Molekylformel C12 H10 O
Molekylvikt (g/mol) 170.21
Synonym Acenaphthene-1-hydroxy
IUPAC-namn 1,2-dihydroacenaphthylen-1-ol
LEDER OC1Cc2cccc3cccc1c23
Specifikationer
Kemiskt namn eller material 1-Acenaphthenol
Analyt- eller komponentnamn 1-Acenaphthenol
Frakt skick Room Temperature
InChI formel InChI=1S/C12H10O/c13-11-7-9-5-1-3-8-4-2-6-10(11)12(8)9/h1-6,11,13H,7H2
Lösningstyp Neat
Industry Type Forensics
Kvantitet 1 g
Formel vikt 170.0732