Läs mer
1-Adamantyl Methyl Ketoxime, TRC
Produktkod. 30316391
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30316391 100 mg
1 options
Produktkod. 30316391 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA220010100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Adamantyl Methyl Ketoxime
CAS 1707-40-0
Molekylformel C12H19NO
Kvantitet 100 mg
Synonym Ethanone, 1-tricyclo[3.3.1.13,7]dec-1-yl-, oxime (9CI, ACI), 1-Tricyclo[3.3.1.13,7]dec-1-ylethanone oxime (ACI), Ketone, 1-adamantyl methyl, oxime (6CI, 7CI, 8CI), 1-Adamantyl methyl ketone oxime, 1-Adamantyl methyl ketoxime
InChI formel InChI=1S/C12H19NO/c1-8(13-14)12-5-9-2-10(6-12)4-11(3-9)7-12/h9-11,14H,2-7H2,1H3/b13-8+
LEDER C\C(=N/O)\C12C[C@@H]3C[C@@H](C[C@@H](C3)C1)C2
IUPAC-namn (NE)-N-[1-(1-adamantyl)ethylidene]hydroxylamine
Molekylvikt (g/mol) 193.29
Formel vikt 193.1467