missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Allyl-3-methylimidazolium chloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
50 g
Description
1-Allyl-3-methylimidazolium chloride is used as an ionic liquid as well as nonderivatizing solvent for cellulose. It acts as a plasticizer for cornstarch and finds application as a solid biopolymer electrolyte. Further, it plays an important role in the preparation of reducing sugar.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Allyl-3-methylimidazolium chloride is used as an ionic liquid as well as nonderivatizing solvent for cellulose. It acts as a plasticizer for cornstarch and finds application as a solid biopolymer electrolyte. Further, it plays an important role in the preparation of reducing sugar.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and bases.
1-Allyl-3-methylimidazolium chloride is used as an ionic liquid as well as nonderivatizing solvent for cellulose. It acts as a plasticizer for cornstarch and finds application as a solid biopolymer electrolyte. Further, it plays an important role in the preparation of reducing sugar.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and bases.
Specifications
Melting Point | 49°C to 51°C |
Quantity | 5 g |
Sensitivity | Moisture sensitive |
Formula Weight | 158.63 |
Percent Purity | 98% |
Chemical Name or Material | 1-Allyl-3-methylimidazolium chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'