Läs mer
1-Alpha-Methyl Dapagliflozin, TRC
Produktkod. 30012909
Change view
Kvantitet:
10 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012909 500 mg
30012038 100 mg
17930365 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012909 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM263010500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 714269-57-5
Molekylformel C22 H27 Cl O7
Molekylvikt (g/mol) 438.9
Synonym Methyl 1-C-[4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl]-α-D-glucopyranoside
IUPAC-namn (2S,3R,4S,5S,6R)-2-[4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl]-6-(hydroxymethyl)-2-methoxyoxane-3,4,5-triol
LEDER CCOc1ccc(Cc2cc(ccc2Cl)[C@]3(OC)O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)cc1
Specifikationer
Kemiskt namn eller material (2S,3R,4S,5S,6R)-2-[4-Chloro-3-(4-ethoxybenzyl)phenyl]-6-(hydroxymethyl)-2-methoxy-tetrahydropyran-3,4,5-triol
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C22H27ClO7/c1-3-29-16-7-4-13(5-8-16)10-14-11-15(6-9-17(14)23)22(28-2)21(27)20(26)19(25)18(12-24)30-22/h4-9,11,18-21,24-27H,3,10,12H2,1-2H3/t18-,19-,20+,21-,22+/m1/s1
Formel vikt 438.1445
Procent renhet >95%